Merck
Prohibitin, an antiproliferative protein, is localized to mitochondria.
FEBS letters (1995-01-30)
E Ikonen, K Fiedler, R G Parton, K Simons
RESUMEN
Prohibitin is a ubiquitously expressed protein with antiproliferative properties. When rat prohibitin tagged with a carboxy-terminal c-Myc epitope was expressed in baby hamster kidney cells the protein was targeted to mitochondria. In immunofluorescence microscopy prohibitin colocalized with a mitochondrial marker E3. Immunoelectron microscopy revealed that prohibitin was associated with the periphery of mitochondria. The amino-terminus of prohibitin shares characteristics of the known mitochondrial import signals, and positioning of the tag at the N-terminus causes accumulation of the protein in the cytoplasm. These findings help to direct functional studies on prohibitin and suggest that a mitochondrial protein may act as a tumor suppressor.