Merck
  • Identification of ETFB as a candidate protein that participates in the mechanoregulation of fibroblast cell number in collagen gel culture.
Identification of ETFB as a candidate protein that participates in the mechanoregulation of fibroblast cell number in collagen gel culture.
Journal of dermatological science (2011-09-10)
Shigenari Hirokawa, Tomomasa Shimanuki, Hiroyuki Kitajima, Yasutomo Nishimori, Makoto Shimosaka
RESUMEN
Fibroblast activation is strongly influenced by mechanical environment in the wound-healing process, especially in fibrosis. Mechanically stressed three-dimensional collagen embedded culture is a useful model representing fibroblasts in morphological as well as biochemical situations encountered during fibrosis. To find key proteins involved in reducing the number of fibroblasts during mechanical stress, we performed two-dimensional gel electrophoresis (2DE)-based differential display and siRNA-based functional screening with collagen gel culture focusing on the differences between attached and detached culture environments. Membrane extracts of fibroblasts from 1 day of attached or detached cultures were subjected to 2DE. We compared protein expression levels and identified the attached-culture-dominant proteins by MALDI-TOF-MS. Next, fibroblasts were transfected with siRNA and embedded in collagen gel. Cell number was counted after 3 days in culture. Eight attached culture dominant proteins were identified with MALDI-TOF-MS. Transfection of siRNA against these proteins demonstrated that electron transfer flavoprotein β subunit (ETFB)-specific siRNA reduced the cell number in the attached culture without a decrease in the detached culture. ETFB participates in the mechanoregulation of fibroblast cell number in collagen gel culture.