Merck
  • Breast Cancer Proteomics - Differences in Protein Expression between Estrogen Receptor-Positive and -Negative Tumors Identified by Tandem Mass Tag Technology.
Breast Cancer Proteomics - Differences in Protein Expression between Estrogen Receptor-Positive and -Negative Tumors Identified by Tandem Mass Tag Technology.
Breast care (Basel, Switzerland) (2010-03-01)
Eugen Ruckhäberle, Thomas Karn, Lars Hanker, Josef Schwarz, Peter Schulz-Knappe, Karsten Kuhn, Gitte Böhm, Stefan Selzer, Neukum Erhard, Knut Engels, Uwe Holtrich, Manfred Kaufmann, Achim Rody
RESUMEN
BACKGROUND: Proteomic analysis has become an effective tool in breast cancer research. In this study, we applied the new gel-free tandem mass tag (TMT) reference method for the first time in breast cancer. MATERIALS AND METHODS: Proteomic analysis was used to compare 10 estrogen receptor (ER)-positive and 10 ER-negative samples. The results of the proteomic approach were validated by Western blot, immunohistochemistry and gene expression analysis. RESULTS: 17 proteins with significant differences in expression were identified. 13 proteins were overexpressed in ER-negative tumors and 4 were overexpressed in ER-positive samples. All these proteins were characterized by relatively high cellular abundance. CONCLUSIONS: Our results demonstrate that the gel-free TMT approach allows the quantification of differences in protein expression levels. Further improvement of the sensitivity by subfractionation of the tissue should allow also the identification of low-abundance proteins and might lead to the use of this method in breast cancer research.