Merck
  • p38 mitogen-activated protein kinase (MAPK) promotes cholesterol ester accumulation in macrophages through inhibition of macroautophagy.
p38 mitogen-activated protein kinase (MAPK) promotes cholesterol ester accumulation in macrophages through inhibition of macroautophagy.
The Journal of biological chemistry (2012-02-23)
Shuang Mei, Haihua Gu, Adam Ward, Xuefeng Yang, Huailan Guo, Ka He, Zhenqi Liu, Wenhong Cao
RESUMEN
p38 MAPK has been strongly implicated in the development of atherosclerosis, but its role in cholesterol ester accumulation in macrophages and formation of foam cells, an early step in the development of atherosclerosis, has not been investigated. We addressed this issue and made some brand new observations. First, elevated intracellular cholesterol level induced by the exposure to LDL-activated p38 MAPK and activation of p38 MAPK with anisomycin increased the ratio of cholesterol esters over free cholesterol, whereas inhibition of p38 MAPK with SB203580 or siRNA reduced the LDL loading-induced intracellular accumulation of free cholesterol and cholesterol esters in macrophages. Second, exposure to LDL cholesterol inhibited autophagy in macrophages, and inhibition of autophagy with 3-methyladenine increased intracellular accumulation of cholesterol (free cholesterol and cholesterol esters), whereas activation of autophagy with rapamycin decreased intracellular accumulation of free cholesterol and cholesterol esters induced by the exposure to LDL cholesterol. Third, LDL cholesterol loading-induced inhibition of autophagy was prevented by blockade of p38 MAPK with SB203580 or siRNA. Neutral cholesterol ester hydrolase was co-localized with autophagosomes. Finally, LDL cholesterol loading and p38 activation suppressed expression of the key autophagy gene, ulk1, in macrophages. Together, our results provide brand new insight about cholesterol ester accumulation in macrophages and foam cell formation.