Merck
  • Monocarboxylate transporter 8 deficiency: altered thyroid morphology and persistent high triiodothyronine/thyroxine ratio after thyroidectomy.
Monocarboxylate transporter 8 deficiency: altered thyroid morphology and persistent high triiodothyronine/thyroxine ratio after thyroidectomy.
European journal of endocrinology (2011-08-05)
Eva K Wirth, Sien-Yi Sheu, Jazmin Chiu-Ugalde, Remy Sapin, Marc O Klein, Ilona Mossbrugger, Leticia Quintanilla-Martinez, Martin Hrabĕ de Angelis, Heiko Krude, Thomas Riebel, Karin Rothe, Josef Köhrle, Kurt W Schmid, Ulrich Schweizer, Annette Grüters
RESUMEN
Thyroid hormone transport across the plasma membrane depends on transmembrane transport proteins, including monocarboxylate transporter 8 (MCT8). Mutations in MCT8 (or SLC16A2) lead to a severe form of X-linked psychomotor retardation, which is characterised by elevated plasma triiodothyronine (T(3)) and low/normal thyroxine (T(4)). MCT8 contributes to hormone release from the thyroid gland. To characterise the potential impact of MCT8-deficiency on thyroid morphology in a patient and in Mct8-deficient mice. Thyroid morphology in a patient carrying the A224V mutation was followed by ultrasound imaging for over 10 years. After thyroidectomy, a histopathological analysis was carried out. The findings were compared with histological analyses of mouse thyroids from the Mct8(-/y) model. We show that an inactivating mutation in MCT8 leads to a unique, progressive thyroid follicular pathology in a patient. After thyroidectomy, histological analysis revealed gross morphological changes, including several hyperplastic nodules, microfollicular areas with stromal fibrosis and a small focus of microfollicular structures with nuclear features reminiscent of papillary thyroid carcinoma (PTC). These findings are supported by an Mct8-null mouse model in which we found massive papillary hyperplasia in 6- to 12-month-old mice and nuclear features consistent with PTC in almost 2-year-old animals. After complete thyroidectomy and substitution with levothyroxine (l-T(4)), the preoperative, inadequately low T(4) and free T(4) remained, while increasing the l-T(4) dosage led to T(3) serum concentrations above the normal range. Our results implicate peripheral deiodination in the peculiar hormonal constellation of MCT8-deficient patients. Other MCT8-deficient patients should be closely monitored for potential thyroid abnormalities.