Merck
  • Overexpression of transcription factor sp2 inhibits epidermal differentiation and increases susceptibility to wound- and carcinogen-induced tumorigenesis.
Overexpression of transcription factor sp2 inhibits epidermal differentiation and increases susceptibility to wound- and carcinogen-induced tumorigenesis.
Cancer research (2010-10-21)
Tae-Hyung Kim, Shannon L Chiera, Keith E Linder, Carol S Trempus, Robert C Smart, Jonathan M Horowitz
RESUMEN
Sp proteins are evolutionarily conserved transcription factors required for the expression of a wide variety of genes that are critical for development and cell cycle progression. Deregulated expression of certain Sp proteins is associated with the formation of a variety of human tumors; however, direct evidence that any given Sp protein is oncogenic has been lacking. Here, we report that Sp2 protein abundance in mice increases in concert with the progression of carcinogen-induced murine squamous cell carcinomas. Transgenic mice specifically overexpressing murine Sp2 in epidermal basal keratinocytes were highly susceptible to wound- and carcinogen-induced papillomagenesis. Transgenic animals that were homozygous rather than hemizygous for the Sp2 transgene exhibited a striking arrest in the epidermal differentiation program, perishing within 2 weeks of birth. Our results directly support the likelihood that Sp2 overexpression occurring in various human cancers has significant functional effect.