Merck
Respiratory Epithelial Cells Can Remember Infection: A Proof-of-Concept Study.
The Journal of infectious diseases (2019-11-05)
Jeanne Bigot, Loic Guillot, Juliette Guitard, Manon Ruffin, Harriet Corvol, Michel Chignard, Christophe Hennequin, Viviane Balloy
RESUMEN
Human bronchial epithelial cells play a key role in airway immune homeostasis. We hypothesized that these sentinel cells can remember a previous contact with pathogen compounds and respond nonspecifically to reinfection, a phenomenon called innate immune memory. We demonstrated that their preexposure to Pseudomonas aeruginosa flagellin modify their inflammatory response to a second, nonrelated stimulus, including live pathogens or lipopolysaccharide. Using histone acetyltransferase and methyltransferase inhibitors, we showed that this phenomenon relied on epigenetic regulation. This report is a major breakthrough in the field of multimicrobial respiratory tract infections, wherein control of inflammatory exacerbations is a major therapeutic issue.