Merck
Catalytic Hydrocyanation of Activated Terminal Alkynes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-09-26)
David Tejedor, Samuel Delgado-Hernández, Lucía Colella, Fernando García-Tellado
RESUMEN
A universal, practical and scalable organocatalytic hydrocyanation manifold to provide β-substituted acrylonitriles bearing an electron-withdrawing functionality has been implemented. The catalytic manifold operates under the reactivity generation principle "a good nucleophile generates a strong base", and it uses 1,4-diazabicyclo[2.2.2]octane (DABCO) as the catalyst, activated terminal alkynes as substrates and acetone cyanohydrin as the cyanide source. The acrylonitriles obtained as E,Z mixtures are straightforwardly resolved by simple flash chromatography delivering the pure isomers in preparative amounts.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethynyl p-tolyl sulfone, 98%