- Phosphorylation of MAP kinases by MAP/ERK involves multiple regions of MAP kinases.
Phosphorylation of MAP kinases by MAP/ERK involves multiple regions of MAP kinases.
The Journal of biological chemistry (1999-06-08)
J L Wilsbacher, E J Goldsmith, M H Cobb
PMID10358048
RESUMEN
Mitogen-activated protein (MAP) kinases are activated with great specificity by MAP/ERK kinases (MEKs). The basis for the specific activation is not understood. In this study chimeras composed of two MAP kinases, extracellular signal-regulated protein kinase 2 and p38, were assayed in vitro for phosphorylation and activation by different MEK isoforms to probe the requirements for productive interaction of MAP kinases with MEKs. Experimental results and modeling support the conclusion that the specificity of MEK/MAP kinase phosphorylation results from multiple contacts, including surfaces in both the N- and C-terminal domains.