Merck
Molecular cloning of a human beta 3-adrenergic receptor cDNA.
FEBS letters (1993-06-14)
J M Lelias, M Kaghad, M Rodriguez, P Chalon, J Bonnin, I Dupre, B Delpech, M Bensaid, G LeFur, P Ferrara
RESUMEN
We report the molecular cloning of a beta 3-adrenergic receptor [beta 3-AR] cDNA from human brown adipose tissue. The cDNA-encoded protein is identical to the previously cloned beta 3-AR but with 6 additional amino acids at the C-terminus. The C-terminus is shared by the beta 3 receptors expressed in human neuroblastoma cells [SK-N-MC] [Mol. Pharmacol. 42 (1992) 964-970]. Furthermore, using a polymerase chain reaction strategy we have cloned and sequenced the beta 3-AR introns. Sequence analysis demonstrates that the human beta 3-AR gene comprises at least 3 exons and 2 introns and that the most abundant beta 3-AR transcripts encode a protein with an exon 3-derived C-terminus. Interestingly, although a similar organization has been found in rodent genes, the rat beta 3-AR transcripts encode a receptor with an exon 2-derived C-terminus.