Merck
Fibroblast growth factor receptors from liver vary in three structural domains.
Science (New York, N.Y.) (1991-02-08)
J Z Hou, M K Kan, K McKeehan, G McBride, P Adams, W L McKeehan
RESUMEN
Changes in heparin-binding fibroblast growth factor gene expression and receptor phenotype occur during liver regeneration and in hepatoma cells. The nucleotide sequence of complementary DNA predicts that three amino-terminal domain motifs, two juxtamembrane motifs, and two intracellular carboxyl-terminal domain motifs combine to form a minimum of 6 and potentially 12 homologous polypeptides that constitute the growth factor receptor family in a single human liver cell population. Amino-terminal variants consisted of two transmembrane molecules that contained three and two immunoglobulin-like disulfide loops, as well as a potential intracellular form of the receptor. The two intracellular juxtamembrane motifs differed in a potential serine-threonine kinase phosphorylation site. One carboxyl-terminal motif was a putative tyrosine kinase that contained potential tyrosine phosphorylation sites. The second carboxyl-terminal motif was probably not a tyrosine kinase and did not exhibit the same candidate carboxyl-terminal tyrosine phosphorylation sites.