Merck
  • Monoclonal anti-mouse laminin antibodies: AL-1 reacts with laminin alpha1 chain, AL-2 with laminin beta1 chain, and AL-4 with the coiled-coil domain of laminin beta1 chain.
Monoclonal anti-mouse laminin antibodies: AL-1 reacts with laminin alpha1 chain, AL-2 with laminin beta1 chain, and AL-4 with the coiled-coil domain of laminin beta1 chain.
Matrix biology : journal of the International Society for Matrix Biology (2006-04-25)
Susanne Schéele, Takako Sasaki, Anna Arnal-Estapé, Madeleine Durbeej, Peter Ekblom
RESUMEN
We analyzed the reactivity of three different commercially available rat monoclonal antibodies raised against mouse laminin-alpha1beta1gamma1 (laminin-111), AL-1, AL-2, and AL-4. Using ELISA assays, Western blot analysis and immunostainings we present refined epitope maps for these three laminin monoclonals. AL-1 reacted, as predicted with laminin alpha1 chain. AL-4 has also been marketed as an alpha1 chain specific probe, but we show here that AL-4 detects mouse laminin beta1 chain, in the distal part of the coiled-coil region. AL-2 was predicted to react with all three chains near the cross-region, but seems to primarily react with laminin beta1 chain.