Merck
Activation of heterotrimeric G-protein signaling by a ras-related protein. Implications for signal integration.
The Journal of biological chemistry (2000-06-07)
M J Cismowski, C Ma, C Ribas, X Xie, M Spruyt, J S Lizano, S M Lanier, E Duzic
RESUMEN
Utilizing a functional screen in the yeast Saccharomyces cerevisiae we identified mammalian proteins that activate heterotrimeric G-protein signaling pathways in a receptor-independent fashion. One of the identified activators, termed AGS1 (for activator of G-protein signaling), is a human Ras-related G-protein that defines a distinct subgroup of the Ras superfamily. Expression of AGS1 in yeast and in mammalian cells results in specific activation of Galpha(i)/Galpha(o) heterotrimeric signaling pathways. In addition, the in vivo and in vitro properties of AGS1 are consistent with it functioning as a direct guanine nucleotide exchange factor for Galpha(i)/Galpha(o). AGS1 thus presents a unique mechanism for signal integration via heterotrimeric G-protein signaling pathways.