Merck
  • Leukemia inhibitory factor, interleukin-6, and their receptors are expressed transiently in the olfactory mucosa after target ablation.
Leukemia inhibitory factor, interleukin-6, and their receptors are expressed transiently in the olfactory mucosa after target ablation.
The Journal of comparative neurology (2001-05-23)
B Nan, M L Getchell, J V Partin, T V Getchell
RESUMEN
Removal of the synaptic targets of olfactory receptor neurons by olfactory bulb ablation results in apoptosis of olfactory receptor neurons and up-regulation of proliferation of their progenitors. This study focuses on the expression of the neuropoietic cytokines leukemia inhibitory factor (LIF) and its receptor (LIFR) and interleukin 6 (IL-6) and its receptor (IL-6R) in intercellular signaling pathways in the olfactory mucosa after target ablation. Olfactory bulbectomy (OBX) resulted in several transient, early-onset, temporally integrated events that were detected immunohistochemically. Macrophages infiltrated the olfactory epithelium (OE) by 16 hours post-OBX. LIF expression was up-regulated transiently at 2 days post-OBX, when up-regulated expression of LIFR also was detected on globose basal cells (GBCs), a subpopulation of which are immediate progenitors of olfactory receptor neurons. GBC proliferation peaked at 3--4 days post-OBX. In the olfactory nerve (ON), LIF-positive and IL-6-positive macrophage infiltration was followed by the transient up-regulation of expression of LIFR, IL-6, and IL-6R in ensheathing cells by 3 days post-OBX. The mRNAs for LIF/LIFR, IL-6/IL-6R, and their common signal-transduction molecule, gp130, in olfactory-nasal mucosa from control mice and from 3-day post-OBX mice were detected with reverse transcriptase-polymerase chain reaction (RT-PCR). Analysis of Northern blot and relative quantitative RT-PCR demonstrated similar temporal patterns of changes in relative mRNA levels for both LIF and IL-6, which were up-regulated by 16 hours post-OBX and peaked at 2--3 days post-OBX. These data indicate that LIF from infiltrating macrophages acts as a mitogen for GBCs and that LIF from infiltrating macrophages and IL-6 from infiltrating macrophages and ensheathing cells act as repair factors in the ON.