Documentos clave
EP0030128958
Eppendorf® Twin tec PCR plates, 384 wells
green polypropylene plate, conical bottom colorless wells, pkg of 300 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom colorless wells
green polypropylene plate
size:
384 wells
well maximum volume:
-45 μm
working volume:
-40 μL
binding type:
low binding surface
feature:
skirt (stackable)
material
conical bottom colorless wells
green polypropylene plate
feature
skirt (stackable)
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128958
technique(s)
PCR: suitable
size
384 wells
well maximum volume
-45 μm
working volume
-40 μL
suitability
suitable for PCR
binding type
low binding surface
General description
The Eppendorf “twin.tec” PCR plates are designed for single-use PCR applications. They include torque-resistant polycarbonate frames and polypropylene wells that help to withstand high temperatures and provide optimum heat transfer. Additionally, the raised rims of the plate further enhance effective sealing by forming a highly secure seal with the sealing material.
Application
“Eppendorf” Twin tec PCR plates are suitable for use in:
  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling
Features and Benefits
  • The PCR plates provide easy sample identification and tracking through its alphanumeric grid-referencing.
  • The raised well rims enable effective sealing while minimizing the risk of cross-contamination.
  • Certified to be free of human DNA, DNase, RNase, and PCR inhibitors.
  • Barcode options are available for convenient sample management.
  • These plates are well-suited for use with most common 384-well thermocyclers and have a maximum well volume of 40 ?L.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)