Merck
LC/MS Analysis of CYP2D6 Inhibitors on Ascentis® Express HILIC
LC/MS Analysis of CYP2D6 Inhibitors on Ascentis® Express HILIC application for HPLC
Materials
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
Imipramine hydrochloride
≥99% (TLC)
Paroxetine hydrochloride
United States Pharmacopeia (USP) Reference Standard
Amitriptyline hydrochloride
Pharmaceutical Secondary Standard; Certified Reference Material
Fluoxetine Hydrochloride
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Ascentis Express HILlC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
5 mM ammonium formate in 2:98 (v/v) water:acetonitrile
flow rate
1.2 mL/min
column temp.
50 °C
detector
ESI(+), m/z 100-400
injection
0.5 μL
Description
Analysis Note
This application demonstrates the suitability of the Ascentis Express HILIC for the analysis of CYP2D6 Inhibitors.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany