HPLC Analysis of Aniline Homologs on Discovery® C18
![HPLC Analysis of Aniline Homologs on Discovery® C18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/111/550/9260f9dd-c1e5-4307-a4a0-df7c9df3df09/800/9260f9dd-c1e5-4307-a4a0-df7c9df3df09.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] methanol; [B] water (60:40, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany