Merck
  • Effects of Osteogenic-Conditioned Medium from Human Periosteum-Derived Cells on Osteoclast Differentiation.
Effects of Osteogenic-Conditioned Medium from Human Periosteum-Derived Cells on Osteoclast Differentiation.
International journal of medical sciences (2017-12-05)
Hyun-Chang Park, Young-Bum Son, Sung-Lim Lee, Gyu-Jin Rho, Young-Hoon Kang, Bong-Wook Park, Sung-Hoon Byun, Sun-Chul Hwang, In-Ae Cho, Yeong-Cheol Cho, Iel-Yong Sung, Dong Kyun Woo, June-Ho Byun
ABSTRACT
Stem/progenitor cell-based regenerative medicine using the osteoblast differentiation of mesenchymal stem cells (MSCs) is regarded as a promising approach for the therapeutic treatment of various bone defects. The effects of the osteogenic differentiation of stem/progenitor cells on osteoclast differentiation may have important implications for use in therapy. However, there is little data regarding the expression of osteoclastogenic proteins during osteoblastic differentiation of human periosteum-derived cells (hPDCs) and whether factors expressed during this process can modulate osteoclastogenesis. In the present study, we measured expression of RANKL in hPDCs undergoing osteoblastic differentiation and found that expression of