Merck
  • Preparation of bioactive soluble human leukemia inhibitory factor from recombinant Escherichia coli using thioredoxin as fusion partner.
Preparation of bioactive soluble human leukemia inhibitory factor from recombinant Escherichia coli using thioredoxin as fusion partner.
Protein expression and purification (2010-04-13)
Magda Tomala, Antonina Lavrentieva, Pierre Moretti, Ursula Rinas, Cornelia Kasper, Frank Stahl, Axel Schambach, Eva Warlich, Ulrich Martin, Tobias Cantz, Thomas Scheper
ABSTRACT
Leukemia inhibitory factor (LIF) is a polyfunctional cytokine with numerous regulatory effects in vivo and in vitro. In stem cell cultures it is the essential media supplement for the maintenance of pluripotency of embryonic and induced pluripotent stem cells. With regard to large scale cultures of these cells, LIF is needed in high quality and quantity and represents the major cost determining factor (90%) of the culture media. In this report, we describe a novel production and purification process for human LIF (hLIF) from recombinant Escherichia coli cultures. hLIF was cloned into pET32b and expressed as soluble protein in fusion with thioredoxin. After purification based on membrane adsorber technology, the fusion protein was cleaved using TEV protease. Released, soluble hLIF was subsequently purified by cation exchange chromatography and successfully tested for its biological activity using suspension cultures of murine embryonic and induced pluripotent stem cells. Our novel protocol for the production of recombinant hLIF is very suitable and effective for the production of poorly soluble proteins through expression in fusion with the solubilizing partner thioredoxin.