Merck
  • Severe oxidative stress in an acute inflammatory demyelinating model in the rhesus monkey.
Severe oxidative stress in an acute inflammatory demyelinating model in the rhesus monkey.
PloS one (2017-11-15)
Jordon Dunham, Reinofke van de Vis, Jan Bauer, Jacqueline Wubben, Nikki van Driel, Jon D Laman, Bert A 't Hart, Yolanda S Kap
ABSTRACT
Oxidative stress is increasingly implicated as a co-factor of tissue injury in inflammatory/demyelinating disorders of the central nervous system (CNS), such as multiple sclerosis (MS). While rodent experimental autoimmune encephalomyelitis (EAE) models diverge from human demyelinating disorders with respect to limited oxidative injury, we observed that in a non-human primate (NHP) model for MS, namely EAE in the common marmoset, key pathological features of the disease were recapitulated, including oxidative tissue injury. Here, we investigated the presence of oxidative injury in another NHP EAE model, i.e. in rhesus macaques, which yields an acute demyelinating disease, which may more closely resemble acute disseminated encephalomyelitis (ADEM) than MS. Rhesus monkey EAE diverges from marmoset EAE by abundant neutrophil recruitment into the CNS and destructive injury to white matter. This difference prompted us to investigate to which extent the oxidative pathway features elicited in MS and marmoset EAE are reflected in the acute rhesus monkey EAE model. The rhesus EAE brain was characterized by widespread demyelination and active lesions containing numerous phagocytic cells and to a lesser extent T cells. We observed induction of the oxidative stress pathway, including injury, with a predilection of p22phox expression in neutrophils and macrophages/microglia. In addition, changes in iron were observed. These results indicate that pathogenic mechanisms in the rhesus EAE model may differ from the marmoset EAE and MS brain due to the neutrophil involvement, but may in the end lead to similar induction of oxidative stress and injury.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse