Merck
  • Initiation of DNA replication requires actin dynamics and formin activity.
Initiation of DNA replication requires actin dynamics and formin activity.
The EMBO journal (2017-10-07)
Nikolaos Parisis, Liliana Krasinska, Bethany Harker, Serge Urbach, Michel Rossignol, Alain Camasses, James Dewar, Nathalie Morin, Daniel Fisher
ABSTRACT
Nuclear actin regulates transcriptional programmes in a manner dependent on its levels and polymerisation state. This dynamics is determined by the balance of nucleocytoplasmic shuttling, formin- and redox-dependent filament polymerisation. Here, using
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PCNA (Ab-1) Mouse mAb (PC10), liquid, clone PC10, Calbiochem®
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid