Merck
  • Strategies for Overcoming Resistance in Tumours Harboring BRAF Mutations.
Strategies for Overcoming Resistance in Tumours Harboring BRAF Mutations.
International journal of molecular sciences (2017-03-12)
Nourah Mohammad Obaid, Karen Bedard, Weei-Yuarn Huang
ABSTRACT
The development of resistance to previously effective treatments has been a challenge for health care providers and a fear for patients undergoing cancer therapy. This is an unfortunately frequent occurrence for patients undergoing targeted therapy for tumours harboring the activating V600E mutation of the