Merck
  • MicroRNA hsa-miR-370-3p suppresses the expression and induction of CYP2D6 by facilitating mRNA degradation.
MicroRNA hsa-miR-370-3p suppresses the expression and induction of CYP2D6 by facilitating mRNA degradation.
Biochemical pharmacology (2017-05-30)
Linjuan Zeng, Yinting Chen, Yong Wang, Li-Rong Yu, Bridgett Knox, Jiwei Chen, Tieliu Shi, Si Chen, Zhen Ren, Lei Guo, Yuanfeng Wu, David Liu, Kaihong Huang, Weida Tong, Dianke Yu, Baitang Ning
ABSTRACT
Cytochrome P450 2D6 (CYP2D6) participates in the metabolism of approximately 20-25% of prescribed drugs. Genetic polymorphisms influence the expression and/or activity of CYP2D6, and inter-individual differences in drug activation and elimination caused by CYP2D6 genetic variants were reported. However, little is known about the potential modulation of CYP2D6 expression by microRNAs (miRNAs). In the current study, by using in silico prediction of the stabilities of miRNA/mRNA complexes, we screened 38 miRNA candidates that may interact with the transcript of CYP2D6. An inverse correlation between the expression of miRNA hsa-miR-370-3p and the expression of CYP2D6 was observed in human liver tissue samples. Electrophoretic mobility shift assays confirmed that hsa-miR-370-3p was able to directly bind to its cognate target within the coding region of the CYP2D6 transcript. The transfection of hsa-miR-370-3p mimics into the HepG2