- Differential effects of anoctamins on intracellular calcium signals.
Differential effects of anoctamins on intracellular calcium signals.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-02-12)
Inês Cabrita, Roberta Benedetto, Ana Fonseca, Podchanart Wanitchakool, Lalida Sirianant, Boris V Skryabin, Laura K Schenk, Hermann Pavenstädt, Rainer Schreiber, Karl Kunzelmann
PMID28183802
ABSTRACT
The Ca