Merck
  • The effects of hydrogen sulfide on platelet-leukocyte aggregation and microvascular thrombolysis.
The effects of hydrogen sulfide on platelet-leukocyte aggregation and microvascular thrombolysis.
Platelets (2016-11-08)
Eberhard Grambow, Christian Leppin, Katja Leppin, Günther Kundt, Ernst Klar, Marcus Frank, Brigitte Vollmar
ABSTRACT
The volatile transmitter hydrogen sulfide (H
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture