Merck
  • Effect of cholesterol loaded cyclodextrin supplementation on tyrosine phosphorylation and apoptosis like changes in frozen thawed Hariana bull spermatozoa.
Effect of cholesterol loaded cyclodextrin supplementation on tyrosine phosphorylation and apoptosis like changes in frozen thawed Hariana bull spermatozoa.
Theriogenology (2017-05-24)
Hanuman P Yadav, Anuj Kumar, Nadeem Shah, Dharmendra S Chauhan, Atul Saxena, Sarvajeet Yadav, Dilip Kumar Swain
ABSTRACT
The beneficial effects of cholesterol loaded cyclodextrin (CLC) addition were evaluated in cryopreserved bull semen. Forty ejaculates were collected from Hariana bulls (n = 4), pooled and divided into 4 aliquots. All the aliquots were initially diluted in to egg yolk tris citrate and supplemented with CLC @ 0.5 mg (Group-II), 1.0 mg (G-III) and 2.0 mg (G-IV) CLC/120 × 10