Merck
Neutrophil stunning by metoprolol reduces infarct size.
Nature communications (2017-04-19)
Jaime García-Prieto, Rocío Villena-Gutiérrez, Mónica Gómez, Esther Bernardo, Andrés Pun-García, Inés García-Lunar, Georgiana Crainiciuc, Rodrigo Fernández-Jiménez, Vinatha Sreeramkumar, Rafael Bourio-Martínez, José M García-Ruiz, Alfonso Serrano Del Valle, David Sanz-Rosa, Gonzalo Pizarro, Antonio Fernández-Ortiz, Andrés Hidalgo, Valentín Fuster, Borja Ibanez
ABSTRACT
The β1-adrenergic-receptor (ADRB1) antagonist metoprolol reduces infarct size in acute myocardial infarction (AMI) patients. The prevailing view has been that metoprolol acts mainly on cardiomyocytes. Here, we demonstrate that metoprolol reduces reperfusion injury by targeting the haematopoietic compartment. Metoprolol inhibits neutrophil migration in an ADRB1-dependent manner. Metoprolol acts during early phases of neutrophil recruitment by impairing structural and functional rearrangements needed for productive engagement of circulating platelets, resulting in erratic intravascular dynamics and blunted inflammation. Depletion of neutrophils, ablation of Adrb1 in haematopoietic cells, or blockade of PSGL-1, the receptor involved in neutrophil-platelet interactions, fully abrogated metoprolol's infarct-limiting effects. The association between neutrophil count and microvascular obstruction is abolished in metoprolol-treated AMI patients. Metoprolol inhibits neutrophil-platelet interactions in AMI patients by targeting neutrophils. Identification of the relevant role of ADRB1 in haematopoietic cells during acute injury and the protective role upon its modulation offers potential for developing new therapeutic strategies.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000