Merck
  • Preclinical amyloid pathology biomarker positivity: effects on tau pathology and neurodegeneration.
Preclinical amyloid pathology biomarker positivity: effects on tau pathology and neurodegeneration.
Translational psychiatry (2017-01-11)
K Höglund, S Kern, A Zettergren, A Börjesson-Hansson, H Zetterberg, I Skoog, K Blennow
ABSTRACT
Brain autopsy and biomarker studies indicate that the pathology of Alzheimer's disease (AD) is initiated at least 10-20 years before clinical symptoms. This provides a window of opportunity to initiate preventive treatment. However, this emphasizes the necessity for biomarkers that identify individuals at risk for developing AD later in life. In this cross-sectional study, originating from three epidemiologic studies in Sweden (n=1428), the objective was to examine whether amyloid pathology, as determined by low cerebrospinal fluid (CSF) concentration of the 42 amino acid form of β-amyloid (Aβ42), is associated with biomarker evidence of other pathological changes in cognitively healthy elderly. A total of 129 patients were included and CSF levels of Aβ42, total tau, tau phosphorylated at threonine 181 (p-tau), neurogranin, VILIP-1, VEGF, FABP3, Aβ40, neurofilament light, MBP, orexin A, BDNF and YKL-40 were measured. Among these healthy elderly, 35.6% (N=46) had CSF Aβ42 levels below 530 pg ml