Merck
  • Early detection of whole body radiation induced microstructural and neuroinflammatory changes in hippocampus: A diffusion tensor imaging and gene expression study.
Early detection of whole body radiation induced microstructural and neuroinflammatory changes in hippocampus: A diffusion tensor imaging and gene expression study.
Journal of neuroscience research (2016-07-21)
Mamta Gupta, Sushanta Kumar Mishra, B S Hemanth Kumar, Subash Khushu, Poonam Rana
ABSTRACT
Ionizing radiation is known to a cause systemic inflammatory response within hours of exposure that may affect the central nervous system (CNS). The present study was carried out to look upon the influence of radiation induced systemic inflammatory response in hippocampus within 24 hr of whole body radiation exposure. A Diffusion Tensor Imaging (DTI) study was conducted in mice exposed to a 5-Gy radiation dose through a