Merck
  • Polyclonal B-cell stimulation by L3T4+ T cells in experimental leishmaniasis.
Polyclonal B-cell stimulation by L3T4+ T cells in experimental leishmaniasis.
Infection and immunity (1988-08-01)
M Lohoff, C Matzner, M Röllinghoff
ABSTRACT
The well-established polyclonal B-cell stimulation in the lymphoid organs in mice infected with Leishmania major is thought to be dependent on T cells. Here we present clear experimental evidence that this is indeed the case by showing that BALB/c-derived, L3T4-positive L. major-specific T cells induce syngeneic B cells to polyclonal proliferation and immunoglobulin production.