Merck
FOXO1 inhibits osteoclastogenesis partially by antagnozing MYC.
Scientific reports (2015-11-17)
Peng Tan, Hanfeng Guan, Linka Xie, Baoguo Mi, Zhong Fang, Jing Li, Feng Li
ABSTRACT
FOXO transcription factors especially FOXO1 have profound roles in bone development and remodeling. The regulation of cells of the osteoblast lineage by FOXOs is suggested to be stage-specific or context dependent. Intriguingly, recent studies on the role played by FOXOs in osteoclastogenesis reached different conclusion. Bartell et al. showed that FOXOs restrained osteoclastogenesis and bone resorption partially by upregulation of the H2O2-inactivating enzyme catalase. Wang et al. demonstrated that FOXO1 activated osteoclast formation. In the present study, we confirmed the results of Bartell et al. that FOXO1 expression was reduced upon stimulation of RANKL; FOXO1 inhibition promoted and FOXO1 activation repressed, osteoclast differentiation and activity; the inhibitory effect of FOXO1 on osteoclastogenesis was partially mediated by ROS since treatment with ROS scavengers cancelled the effect of FOXO1 inhibition on osteoclastogenesis. We further investigated the mechanisms responsible for repressed osteoclastogenesis by FOXO1. We found that FOXO1 inhibition modulated MAPKs, NF-κB and AP-1. Finally, we proved that the inhibitory effect of FOXO1 on osteoclast formation was partially mediated by MYC suppression by showing that MYC repression almost totally abrogated the effect of FOXO1 inhibition on osteoclastogenesis. To conclude, our study confirmed FOXO1 as a cell-autonomous inhibitor of osteoclastogenesis.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
10058-F4, ≥98% (HPLC), solid
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)