Merck
  • Decreased bone marrow stromal cells activity involves in unilateral anterior crossbite-induced early subchondral bone loss of temporomandibular joints.
Decreased bone marrow stromal cells activity involves in unilateral anterior crossbite-induced early subchondral bone loss of temporomandibular joints.
Archives of oral biology (2014-06-16)
Ting Yang, Jing Zhang, Yukun Cao, Mian Zhang, Lei Jing, Kai Jiao, Shibin Yu, Meiqing Wang
ABSTRACT
Subchondral bone loss in mandibular condyles was reported to be induced by experimentally created unilateral anterior crossbite (UAC) which altered the occlusal load distribution and hereafter the temporomandibular joint (TMJ) remodelling process. However, the initial cellular responses are poorly understood. In the present study, changes in osteoblast and osteoclast activities in TMJ subchondral bone were investigated using the rats treated with UAC. Forty rats were randomly divided into UAC and control groups, and sampled at 2 weeks after the operation. Subchondral bone loss was evaluated by micro-CT. Osteoclast and osteoblast activities were analyzed by real-time PCR. The osteoblast differentiation of the locally isolated BMSCs from TMJ subchondral bone was assessed by Alizarin red staining. The migration of BMSCs was detected by transwell assays. Compared with the age-matched controls, TMJ subchondral bone loss was observed in the UAC-treated rats (p<0.05). The osteoblast activity evaluated by real-time PCR and osteoblast number revealed by immunohistochemical staining were reduced in the TMJ subchondral bone of UAC rats (p<0.05), and the capability of proliferation, migration and osteoblast differentiation were all decreased in the locally isolated BMSCs from the UAC group (p<0.05). The present data demonstrated an involvement of reduced BMSCs activity in the initiation of the mandibular subchondral bone loss at the early stage of installation of the aberrant prostheses.
Product Description
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated