Merck
CTLA-4 (CD152) enhances the Tc17 differentiation program.
European journal of immunology (2014-04-12)
Jonas Pick, Aditya Arra, Holger Lingel, J Kolja Hegel, Magdalena Huber, Gopala Nishanth, Gerhard Jorch, Klaus-Dieter Fischer, Dirk Schlüter, Kerry Tedford, Monika C Brunner-Weinzierl
ABSTRACT
Although CD8(+) T cells that produce IL-17 (Tc17 cells) have been linked to host defense, Tc17 cells show reduced cytotoxic activity, which is the characteristic function of CD8(+) T cells. Here, we show that CTLA-4 enhances the frequency of IL-17 in CD8(+) T cells, indicating that CTLA-4 (CD152) specifically promotes Tc17 differentiation. Simultaneous stimulation of CTLA-4(+/+) and CTLA-4(-/-) T cells in cocultures and agonistic CTLA-4 stimulation unambiguously revealed a cell-intrinsic mechanism for IL-17 control by CTLA-4. The quality of CTLA-4-induced Tc17 cells was tested in vivo, utilizing infection with the facultative intracellular bacterium Listeria monocytogenes (LM). Unlike CTLA-4(+/+) Tc17 cells, CTLA-4(-/-) were nearly as efficient as Tc1 CTLA-4(+/+) cells in LM clearance. Additionally, adoptively transferred CTLA-4(-/-) Tc17 cells expressed granzyme B after rechallenge, and produced Tc1 cytokines such as IFN-γ and TNF-α, which strongly correlate with bacterial clearance. CTLA-4(+/+) Tc17 cells demonstrated a high-quality Tc17 differentiation program ex vivo, which was also evident in isolated IL-17-secreting Tc17 cells, with CTLA-4-mediated enhanced upregulation of Tc17-related molecules such as IL-17A, RORγt, and IRF-4. Our results show that CTLA-4 promotes Tc17 differentiation that results in robust Tc17 responses. Its inactivation might therefore represent a central therapeutic target to enhance clearance of infection.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O