Merck
  • The effect of cloudy apple juice on hepatic and mammary gland phase I and II enzymes induced by DMBA in female Sprague-Dawley rats.
The effect of cloudy apple juice on hepatic and mammary gland phase I and II enzymes induced by DMBA in female Sprague-Dawley rats.
Drug and chemical toxicology (2014-03-04)
Hanna Szaefer, Violetta Krajka-Kuźniak, Ewa Ignatowicz, Teresa Adamska, Jarosław Markowski, Wanda Baer-Dubowska
ABSTRACT
Apples abundant in phenolic compounds show a variety of biological activities that may contribute to health beneficial effects against cardiovascular diseases, diabetes, obesity and cancer. We investigated the effect of cloudy apple juice (CAJ) on the hepatic and mammary gland carcinogen metabolizing enzymes, DNA damage and liver injury, altered by 7,12-dimethylbenz[a]anthracene (DMBA). Sprague-Dawley female rats were gavaged with CAJ (10 ml/kg b.w.) for 28 consecutive days. DMBA was administered i.p. on the 27th and the 28th days. In the liver, feeding with CAJ decreased the activities of CYP1A1 and 1A2 and increased phase II enzymes. The activities of all enzymes tested were enhanced in the animals treated with DMBA alone and in combination with CAJ. The most significant changes in the level of the hepatic enzymes tested were observed for GST alpha and NQO1. In mammary gland CAJ induced an increase in the level of GST mu and GST pi, while DMBA and CAJ combined administration elevated GST pi only. This may be beneficial as GST pi is involved in the DMBA detoxification. Additionally, pretreatment with CAJ reduced the level of most of the blood biochemical liver and kidney markers elevated as a result of DMBA treatment. These findings indicate that CAJ may interfere with enzyme system involved in carcinogen metabolism. However, this effect seems to be dependent on tissue and carcinogen and is moderately effective in the case of DMBA. Moreover, CAJ can also provide some protection against the liver and kidney damage.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Resorufin ethyl ether, ≥98% (TLC), powder
Sigma-Aldrich
3,3′-Methylene-bis(4-hydroxycoumarin)
Sigma-Aldrich
Resorufin ethyl ether, suitable for fluorescence, ≥95% (UV)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium 2,6-dichloroindophenolate hydrate, ACS reagent
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Resorufin, 95%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)