Merck
  • Functional implications of calcium permeability of the channel formed by pannexin 1.
Functional implications of calcium permeability of the channel formed by pannexin 1.
The Journal of cell biology (2006-08-16)
Fabien Vanden Abeele, Gabriel Bidaux, Dmitri Gordienko, Benjamin Beck, Yuri V Panchin, Ancha V Baranova, Dmitry V Ivanov, Roman Skryma, Natalia Prevarskaya
ABSTRACT
Although human pannexins (PanX) are homologous to gap junction molecules, their physiological function in vertebrates remains poorly understood. Our results demonstrate that overexpression of PanX1 results in the formation of Ca(2+)-permeable gap junction channels between adjacent cells, thus, allowing direct intercellular Ca(2+) diffusion and facilitating intercellular Ca(2+) wave propagation. More intriguingly, our results strongly suggest that PanX1 may also form Ca(2+)-permeable channels in the endoplasmic reticulum (ER). These channels contribute to the ER Ca(2+) leak and thereby affect the ER Ca(2+) load. Because leakage remains the most enigmatic of those processes involved in intracellular calcium homeostasis, and the molecular nature of the leak channels is as yet unknown, the results of this work provide new insight into calcium signaling mechanisms. These results imply that for vertebrates, a new protein family, referred to as pannexins, may not simply duplicate the connexin function but may also provide additional pathways for intra- and intercellular calcium signaling and homeostasis.