Merck
  • Cyclic AMP stimulates somatostatin gene transcription by phosphorylation of CREB at serine 133.
Cyclic AMP stimulates somatostatin gene transcription by phosphorylation of CREB at serine 133.
Cell (1989-11-17)
G A Gonzalez, M R Montminy
ABSTRACT
In this paper, we demonstrate that phosphorylation of CREB at Ser-133 is induced 6-fold in vivo, following treatment of PC12 cells with forskolin. By contrast, no such induction was observed in the kinase A-deficient PC12 line A126-1B2 (A126). Using F9 teratocarcinoma cells, which are unresponsive to cAMP, we initiated a series of transient expression experiments to establish a causal link between phosphorylation of CREB and trans-activation of cAMP-responsive genes. Inactivating the kinase A phosphorylation site by in vitro mutagenesis of the cloned CREB cDNA at Ser-133 completely abolished CREB transcriptional activity. As CREB mutants containing acidic residues in place of the Ser-133 phosphoacceptor were also transcriptionally inactive, these results suggest that phosphorylation of CREB may stimulate transcription by a mechanism other than by simply providing negative charge.