Merck
  • Expression of organic anion-transporting polypeptides 1B3, 1B1, and 1A2 in human pancreatic cancer reveals a new class of potential therapeutic targets.
Expression of organic anion-transporting polypeptides 1B3, 1B1, and 1A2 in human pancreatic cancer reveals a new class of potential therapeutic targets.
OncoTargets and therapy (2011-05-10)
Valentinos Kounnis, Elli Ioachim, Martin Svoboda, Andreas Tzakos, Ioannis Sainis, Theresia Thalhammer, Georg Steiner, Evangelos Briasoulis
ABSTRACT
Organic anion-transporting polypeptides (OATPs) are influx transporters that mediate intracellular uptake of selective endogenous and xenobiotic compounds. Identification of new molecular targets and discovery of novel targeted therapies is top priority for pancreatic cancer, which lacks any effective therapy. We studied expression of OATP 1A2, 1B1, and 1B3 in pancreatic cancer tissue and in cell lines. Formalin-fixed paraffin-embedded biopsy material of 12 human pancreatic cancers was immunohistochemically assessed for protein expression of the three studied influx transporters. Immunohistochemistry was evaluated by experienced pathologists and quantified by use of an automated image analysis system. BxPC-3 and MIA PaCa-2 pancreatic cancer cell lines were used to quantify transcripts of OATP 1B1 and 1B3. OATP 1A2, 1B1, and 1B3 proteins were found ubiquitously expressed in all studied cases. Quantification performed by HistoQuest system revealed that mean intensity was 53 for 1A2, 45 for 1B1, and 167 for OATP 1B1/1B3 on a range scale 0-250 units. At mRNA level, 1B1 and 1B3 were overexpressed in both studied cancer cell lines but not in normal pancreatic tissue. OATPs 1A2, 1B1, and 1B3 are highly expressed in pancreatic adenocarcinoma. We suggest that expression of these transporters in pancreatic cancer justify research efforts towards discovery of novel therapeutics targeting OATPs.