Merck
  • The zinc finger protein Gsf1 regulates Gsf2-dependent flocculation in fission yeast.
The zinc finger protein Gsf1 regulates Gsf2-dependent flocculation in fission yeast.
FEMS yeast research (2013-01-15)
Tomohiko Matsuzawa, Youko Kageyama, Kazuyoshi Ooishi, Makoto Kawamukai, Kaoru Takegawa
ABSTRACT
Fission yeast flocculates nonsexually by induction of the flocculin encoded by gsf2(+) which is controlled by the positive regulator Mbx2. Here, we report a novel gene designated gsf1(+) found to be a negative regulator of nonsexual flocculation. We identified gsf1(+) as a multicopy suppressor of a sam2 mutation, which caused growth sensitivity to Ca(2+) and also found a nonsense mutation in gsf1(+) in a previously isolated gsf1 mutant. The gsf1(+) gene encodes a 547-aa protein containing a Zn(2)-Cys(6) binuclear cluster-type zinc finger motif. The Gsf1 protein localized in the nucleus, consistent with a role as a transcription factor. Deletion of gsf1(+) resulted in nonsexual flocculation inducible by CaCl2 , which was suppressed by the addition of EDTA or galactose. Both gsf2(+) and mbx2(+) were highly expressed in the gsf1 mutant. gsf1∆ gsf2∆ and gsf1∆ mbx2∆ double mutants did not flocculate, suggesting that gsf1(+) is an upstream regulator. In addition, the gsf1 mutant was sensitive to CaCl2 , KCl, HU, and TBZ, consistent with the possibility that gsf1(+) plays a role in functions unrelated to flocculation. Taken together, these results suggest that nonsexual flocculation in fission yeast is negatively controlled by Gsf1, which controls expression of mbx2(+) and gsf2(+) .
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%