Merck
  • Inhibition of hepatitis C virus NS5B polymerase by S-trityl-L-cysteine derivatives.
Inhibition of hepatitis C virus NS5B polymerase by S-trityl-L-cysteine derivatives.
European journal of medicinal chemistry (2012-01-28)
Daniel B Nichols, Guy Fournet, K R Gurukumar, Amartya Basu, Jin-Ching Lee, Naoya Sakamoto, Frank Kozielski, Ira Musmuca, Benoît Joseph, Rino Ragno, Neerja Kaushik-Basu
ABSTRACT
Structure-based studies led to the identification of a constrained derivative of S-trityl-l-cysteine (STLC) scaffold as a candidate inhibitor of hepatitis C virus (HCV) NS5B polymerase. A panel of STLC derivatives were synthesized and investigated for their activity against HCV NS5B. Three STLC derivatives, 9, F-3070, and F-3065, were identified as modest HCV NS5B inhibitors with IC(50) values between 22.3 and 39.7 μM. F-3070 and F-3065 displayed potent inhibition of intracellular NS5B activity in the BHK-NS5B-FRLuc reporter and also inhibited HCV RNA replication in the Huh7/Rep-Feo1b reporter system. Binding mode investigations suggested that the STLC scaffold can be used to develop new NS5B inhibitors by further chemical modification at one of the trityl phenyl group.
Sigma-Aldrich
(+)-S-Trityl-L-cysteine, 97%
Pricing and availability is not currently available.