Merck
Mass spectrometry-based quantification of pseudouridine in RNA.
Journal of the American Society for Mass Spectrometry (2011-09-29)
Balasubrahmanyam Addepalli, Patrick A Limbach
ABSTRACT
Direct detection of pseudouridine (ψ), an isomer of uridine, in RNA is challenging. The most popular method requires chemical derivatization using N-cyclohexyl-N'-β-(4-methylmorpholinum ethyl) carbodiimide p-tosylate (CMCT) followed by radiolabeled primer extension mediated by reverse transcriptase. More recently, mass spectrometry (MS)-based approaches for sequence placement of pseudouridine in RNA have been developed. Nearly all of these approaches, however, only yield qualitative information regarding the presence or absence of pseudouridine in a given RNA population. Here, we have extended a previously developed liquid chromatography tandem mass spectrometry (LC-MS/MS) method to enable both the qualitative and quantitative analysis of pseudouridine. Quantitative selected reaction monitoring (SRM) assays were developed using synthetic oligonucleotides, with or without pseudouridine, and the results yielded a linear relationship between the ion abundance of the pseudouridine-specific fragment ion and the amount of pseudouridine-containing oligonucleotide present in the original sample. Using this quantitative SRM assay, the extent of pseudouridine hypomodification in the conserved T-loop of tRNA isolated from two different Escherichia coli strains was established.
Sigma-Aldrich
N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide methyl-p-toluenesulfonate, 95%