Merck
  • 7,7''-Dimethoxyagastisflavone-induced apoptotic or autophagic cell death in different cancer cells.
7,7''-Dimethoxyagastisflavone-induced apoptotic or autophagic cell death in different cancer cells.
Phytotherapy research : PTR (2011-09-15)
Chia-Hsiang Hwang, Yu-Ling Lin, Yen-Ku Liu, Chia-Hung Chen, Hsin-Yi Wu, Cheng-Chang Chang, Chao-Yuan Chang, Yu-Kuo Chang, Yi-Han Chiu, Kuang-Wen Liao, Yiu-Kay Lai
ABSTRACT
7,7''-Dimethoxyagastisflavone (DMGF), a biflavonoid isolated from the needles of Taxus × media cv. Hicksii, was evaluated for its antiproliferative and antineoplastic effects in three human cancer cell lines. Interestingly, DMGF caused cell death via different pathways in different cancer cells. DMGF induced apoptosis, activated caspase-3 activity and changed the mitochondrial membrane potential in HT-29 human colon cancer cells. However, the apoptotic pathway is not the major pathway involved in DMGF-induced cell death in A549 human lung cancer cells and HepG2 human hepatoma cells. Treatment with 3-MA, an inhibitor of autophagy, significantly decreased DMGF-induced cell death in HepG2 and A549 cells, but did not affect DMGF-induced cell death in HT-29 cells. Following DMGF treatment, the HepG2 cells increased expression of LC3B-II, a marker used to monitor autophagy in cells. Thus, DMGF induced apoptotic cell death in HT-29 cells, triggered both apoptotic and autophagic death in A549 cells and induced autophagic cell death in HepG2 cells.
Sigma-Aldrich
Baccatin III, ≥95% (HPLC)