Merck
  • Expression of neurotransmitter genes in motor regions of the dyskinetic rat after iminodipropionitrile.
Expression of neurotransmitter genes in motor regions of the dyskinetic rat after iminodipropionitrile.
Neuroscience letters (2003-07-17)
J A Fritschi, Th Lauterburg, J-M Burgunder
ABSTRACT
Rats treated with iminodipropionitrile develop a neurobehaviour syndrome with dyskinesia. Searching for the molecular correlates, we have examined the expression of selected genes involved in neurotransmission in motor regions using hybridization histochemistry. Frontal cortical and thalamic vasoactive intestinal peptide (VIP) expression, and striatal dynorphin, enkephalin (ENK) and substance P expression were increased. No change in cortical cholecystokinin (CCK), ENK, glutamic acid decarboxylase (GAD) and somatostatin (SRIF) expression, in striatal GAD, SRIF, nitric oxide synthase (NOS) and guanylate cyclase expression, and in thalamic CCK, GAD and thyrotropin-releasing hormone expression was found. NOS expression in the subthalamic nucleus as well as tyrosine hydroxylase, GAD and CCK expression in the substantia nigra were unchanged. These results confirm the involvement of striatal projection neurons in dyskinesia and suggest a novel role for VIP.
Sigma-Aldrich
3,3′-Iminodipropionitrile, technical grade, 90%