Merck
  • Alternative pre-mRNA processing regulates cell-type specific expression of the IL4l1 and NUP62 genes.
Alternative pre-mRNA processing regulates cell-type specific expression of the IL4l1 and NUP62 genes.
BMC biology (2005-07-21)
Stefan Wiemann, Anja Kolb-Kokocinski, Annemarie Poustka
ABSTRACT
Given the complexity of higher organisms, the number of genes encoded by their genomes is surprisingly small. Tissue specific regulation of expression and splicing are major factors enhancing the number of the encoded products. Commonly these mechanisms are intragenic and affect only one gene. Here we provide evidence that the IL4I1 gene is specifically transcribed from the apparent promoter of the upstream NUP62 gene, and that the first two exons of NUP62 are also contained in the novel IL4I1_2 variant. While expression of IL4I1 driven from its previously described promoter is found mostly in B cells, the expression driven by the NUP62 promoter is restricted to cells in testis (Sertoli cells) and in the brain (e.g., Purkinje cells). Since NUP62 is itself ubiquitously expressed, the IL4I1_2 variant likely derives from cell type specific alternative pre-mRNA processing. Comparative genomics suggest that the promoter upstream of the NUP62 gene originally belonged to the IL4I1 gene and was later acquired by NUP62 via insertion of a retroposon. Since both genes are apparently essential, the promoter had to serve two genes afterwards. Expression of the IL4I1 gene from the "NUP62" promoter and the tissue specific involvement of the pre-mRNA processing machinery to regulate expression of two unrelated proteins indicate a novel mechanism of gene regulation.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type IV, ≥4.0 units/mg protein, aqueous suspension
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type I (dried venom)
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus atrox (Western Diamondback Rattlesnake), Type VI, dried venom