Merck
  • Beta2-adrenoreceptor agonist clenbuterol produces transient decreases in alpha-synuclein mRNA but no long-term reduction in protein.
Beta2-adrenoreceptor agonist clenbuterol produces transient decreases in alpha-synuclein mRNA but no long-term reduction in protein.
NPJ Parkinson's disease (2022-05-25)
Joseph R Patterson, Warren D Hirst, Jacob W Howe, Christopher P Russell, Allyson Cole-Strauss, Christopher J Kemp, Megan F Duffy, Jared Lamp, Andrew Umstead, Michael Kubik, Anna C Stoll, Irving E Vega, Kathy Steece-Collier, Yi Chen, Anne C Campbell, Catherine L Nezich, Kelly E Glajch, Caryl E Sortwell
ABSTRACT
β2-adrenoreceptor (β2AR) agonists have been associated with a decreased risk of developing Parkinson's disease (PD) and are hypothesized to decrease expression of both alpha-synuclein mRNA (Snca) and protein (α-syn). Effects of β2AR agonist clenbuterol on the levels of Snca mRNA and α-syn protein were evaluated in vivo (rats and mice) and in rat primary cortical neurons by two independent laboratories. A modest decrease in Snca mRNA in the substantia nigra was observed after a single acute dose of clenbuterol in rats, however, this decrease was not maintained after multiple doses. In contrast, α-syn protein levels remained unchanged in both single and multiple dosing paradigms. Furthermore, clenbuterol did not decrease Snca in cultured rat primary cortical neurons, or decrease Snca or α-syn in mice. Additionally, compared to the single-dose paradigm, repeat dosing resulted in substantially lower levels of clenbuterol in plasma and brain tissue in rodents. Based on our observations of a transient decrease in Snca and no effect on α-syn protein in this preclinical study, these data support the conclusion that clenbuterol is not likely a viable disease-modifying strategy for PD.
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®