Merck
  • Possible role of NF-ĸB signals in heat stress-associated increase in protein content of cultured C2C12 cells.
Possible role of NF-ĸB signals in heat stress-associated increase in protein content of cultured C2C12 cells.
Cells, tissues, organs (2011-03-23)
Yoshitaka Ohno, Sumio Yamada, Takao Sugiura, Yoshinobu Ohira, Toshitada Yoshioka, Katsumasa Goto
ABSTRACT
Heat stress is one of the hypertrophic stimuli on mammalian skeletal muscle. Nuclear factor-κB (NF-κB) signaling plays an important role in the regulation of skeletal muscle mass. However, the effects of heat stress on NF-κB signaling in skeletal muscle cells remain unclear. Effects of heat stress and/or administration of BAY11-7082, an inhibitor of NF-κB, on NF-κB signals and protein content of skeletal muscle were studied by using cell culture system. Differentiated mouse myoblasts (C2C12) were subjected to either (1) control (cultured at 37°C without BAY11-7082), (2) heat stress at 41°C for 60 min, (3) BAY11-7082 administration (1.25 μM) or (4) heat stress combined with BAY11-7082 administration. Heat shock protein 72 (HSP72) was upregulated by heat stress with or without administration of BAY11-7082. The increase in inhibitor of κBα (IκBα), which regulates the phosphorylation of NF-κB, and the decrease in phosphorylated NF-κB were also induced by administration of BAY11-7082 and/or heat stress. Protein content in C2C12 cells was increased by the administration of BAY11-7082 with a semi-logarithm fashion. Significant increases in the protein content of C2C12 cells were observed 48 h following heating with or without administration of BAY11-7082. These observations suggest that heat stress might increase muscle protein through the downregulation of NF-κB signaling. Inhibition of NF-κB induced by application of heat stress might be one of the hypertrophic stimuli on skeletal muscle cells.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution