Merck
  • Dietary Magnesium Insufficiency Induces Salt-Sensitive Hypertension in Mice Associated With Reduced Kidney Catechol-O-Methyl Transferase Activity.
Dietary Magnesium Insufficiency Induces Salt-Sensitive Hypertension in Mice Associated With Reduced Kidney Catechol-O-Methyl Transferase Activity.
Hypertension (Dallas, Tex. : 1979) (2021-04-13)
Asako Kumagai, Satoru Takeda, Eisei Sohara, Shinichi Uchida, Hiroshi Iijima, Astuo Itakura, Daisuke Koya, Keizo Kanasaki
ABSTRACT
[Figure: see text].