Merck
  • Primary glomus tumour of the pituitary gland: diagnostic challenges of a rare and potentially aggressive neoplasm.
Primary glomus tumour of the pituitary gland: diagnostic challenges of a rare and potentially aggressive neoplasm.
Virchows Archiv : an international journal of pathology (2020-09-13)
Boon Leong Quah, Carmine Antonio Donofrio, Stefano La Rosa, Jean-Philippe Brouland, Giulia Cossu, Ibrahim Djoukhadar, Helen Mayers, Patrick Shenjere, Marta Pereira, Omar N Pathmanaban, Muhammed O Murtaza, Rao Gattamaneni, Federico Roncaroli, Konstantina Karabatsou
ABSTRACT
Primary non-neuroendocrine tumours of the pituitary gland and sella are rare lesions often challenging to diagnose. We describe two cases of clinically aggressive primary glomus tumour of the pituitary gland. The lesions occurred in a 63-year-old male and a 30-year-old female who presented with headache, blurred vision and hypopituitarism. Neuroimaging demonstrated large sellar and suprasellar tumours invading the surrounding structures. Histologically, the lesions were characterised by angiocentric sheets and nests of atypical cells that expressed vimentin, smooth muscle actin and CD34. Perivascular deposition of collagen IV was also a feature. Case 2 expressed synaptophysin. INI-1 (SMARCB1) expression was preserved. Both lesions were mitotically active and demonstrated a Ki-67 labelling index of 30%. Next-generation sequencing performed in case 1 showed no mutations in the reading frame of 37 commonly mutated oncogenes, including BRAF and KRAS. Four pituitary glomus tumours have previously been reported, none of which showed features of malignant glomus tumour. Similar to our two patients, three previous examples displayed aggressive behaviour.