Merck
  • Vesicular nucleotide transporter-immunoreactive type I cells associated with P2X3-immunoreactive nerve endings in the rat carotid body.
Vesicular nucleotide transporter-immunoreactive type I cells associated with P2X3-immunoreactive nerve endings in the rat carotid body.
The Journal of comparative neurology (2019-12-07)
Takuya Yokoyama, Yoshio Yamamoto, Masato Hirakawa, Kouki Kato, Tomoyuki Saino
ABSTRACT
ATP is the major excitatory transmitter from chemoreceptor type I cells to sensory nerve endings in the carotid body, and has been suggested to be released by exocytosis from these cells. We investigated the mRNA expression and immunohistochemical localization of vesicular nucleotide transporter (VNUT) in the rat carotid body. RT-PCR detected mRNA expression of VNUT in extracts of the tissue. Immunoreactivity for VNUT was localized in a part of type I cells immunoreactive for synaptophysin (SYN), but not in glial-like type II cells immunoreactive for S100 and S100B. Among SYN-immunoreactive type I cells, VNUT immunoreactivity was selectively localized in the sub-population of tyrosine hydroxylase (TH)-immunorective type I cells associated with nerve endings immunoreactive for the P2X3 purinoceptor; however, it was not detected in the sub-population of type I cells immunoreactive for dopamine beta-hydroxylase. Multi-immunolabeling for VNUT, P2X3, and Bassoon revealed that Bassoon-immunoreactive products were localized in type I cells with VNUT immunoreactivity, and accumulated on the contact side of P2X3-immunoreactive nerve endings. These results revealed the selective localization of VNUT in the subpopulation of TH-immunoreactive type I cells attached to sensory nerve endings and suggested that these cells release ATP by exocytosis for chemosensory transmission in the carotid body.
Sigma-Aldrich
Anti-Vesicular Nucleotide Transporter (VNUT Antibody), serum, from guinea pig
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Anti-Dopamine β Hydroxylase Antibody, clone 4F10.2, ascites fluid, clone 4F10.2, Chemicon®
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid