Merck
  • Role of JAK-STAT signaling in the pathogenic behavior of fibroblast-like synoviocytes in rheumatoid arthritis: Effect of the novel JAK inhibitor peficitinib.
Role of JAK-STAT signaling in the pathogenic behavior of fibroblast-like synoviocytes in rheumatoid arthritis: Effect of the novel JAK inhibitor peficitinib.
European journal of pharmacology (2020-06-21)
Takashi Emori, Michiko Kasahara, Shingo Sugahara, Motomu Hashimoto, Hiromu Ito, Shuh Narumiya, Yasuyuki Higashi, Yasutomo Fujii
ABSTRACT
Rheumatoid arthritis (RA) fibroblast-like synoviocytes (RA-FLS) play a crucial role in the pathogenesis of RA. RA-FLS display passive pro-inflammatory responses and self-directed aggressive responses, such as pro-inflammatory mediator production, reduced apoptosis and formation of a thickened synovial lining. Evidence suggests a role for Janus kinase (JAK)-signal transducer and transcriptional activator (STAT) signaling in the passive response but the aggressive behavior of RA-FLS is poorly understood. The pharmacologic effects of the novel JAK inhibitor, peficitinib, on cytokine-induced intracellular signaling and self-directed aggressive behavior of RA-FLS (e.g., increased expression of apoptosis-resistant genes and sodium nitroprusside-induced apoptosis) were investigated and compared with approved JAK inhibitors. RA-FLS assembly to form a lining-like structure and pro-inflammatory mediator production was investigated in three-dimensional (3D)-micromass culture. Peficitinib inhibited STAT3 phosphorylation in RA-FLS following induction by interferon (IFN)-α2b, IFN-γ, interleukin (IL)-6, oncostatin M, and leukemia inhibitory factor in a concentration-related manner, and was comparable to approved JAK inhibitors, tofacitinib and baricitinib. Peficitinib and tofacitinib suppressed autocrine phosphorylation of STAT3 and expression of apoptosis-resistant genes, and promoted cell death. In 3D-micromass culture, peficitinib reduced multi-layered RA-FLS cells to a thin monolayer, an effect less pronounced with tofacitinib. Both compounds attenuated production of vascular endothelial growth factor-A, matrix metalloproteinases, IL-6 and tumor necrosis factor superfamily-11. This study confirmed the pathogenic role of uncontrolled JAK-STAT signaling in the aggressive and passive responses of RA-FLS that are critical for RA progression. The novel JAK inhibitor peficitinib suppressed the pro-inflammatory behavior of RA-FLS, accelerated cell death and abrogated thickening of the synovium.