Merck
  • Alterations of gut microbiota contribute to the progression of unruptured intracranial aneurysms.
Alterations of gut microbiota contribute to the progression of unruptured intracranial aneurysms.
Nature communications (2020-06-27)
Hao Li, Haochen Xu, Youxiang Li, Yuhua Jiang, Yamin Hu, Tingting Liu, Xueqing Tian, Xihai Zhao, Yandong Zhu, Shuxia Wang, Chunrui Zhang, Jing Ge, Xuliang Wang, Hongyan Wen, Congxia Bai, Yingying Sun, Li Song, Yinhui Zhang, Rutai Hui, Jun Cai, Jingzhou Chen
ABSTRACT
Unruptured intracranial aneurysm (UIA) is a life-threatening cerebrovascular condition. Whether changes in gut microbial composition participate in the development of UIAs remains largely unknown. We perform a case-control metagenome-wide association study in two cohorts of Chinese UIA patients and control individuals and mice that receive fecal transplants from human donors. After fecal transplantation, the UIA microbiota is sufficient to induce UIAs in mice. We identify UIA-associated gut microbial species link to changes in circulating taurine. Specifically, the abundance of Hungatella hathewayi is markedly decreased and positively correlated with the circulating taurine concentration in both humans and mice. Consistently, gavage with H. hathewayi normalizes the taurine levels in serum and protects mice against the formation and rupture of intracranial aneurysms. Taurine supplementation also reverses the progression of intracranial aneurysms. Our findings provide insights into a potential role of H. hathewayi-associated taurine depletion as a key factor in the pathogenesis of UIAs.
Sigma-Aldrich
Elastase from porcine pancreas, lyophilized powder, suitable for cell culture